• USA Home
  • Search Results

  • monensinsodiumsalthydrate69285anhydrousbasis22373780anhydrous11

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Monensin sodium salt hydrate

1 Products
Image of Monensin sodium salt hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C36H61NaO11 · xH2O
Molecular Weight:
 692.85 (anhydrous basis)
 E Number:
 E714
 EC Number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More