• USA Home
  • Search Results

  • monobasicpotassiumphosphate13609777877011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Monobasic potassium phosphate

2 Products
Synonym:   Monopotassium phosphate, Potassium dihydrogen phosphate, Potassium phosphate monobasic, prim.-Potassium phosphate
Image of Monobasic potassium phosphate
CAS Number:
Linear Formula:
 KH2PO4
Molecular Weight:
 136.09
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  pK-Values
  Application(s)
Show More