• USA Home
  • Search Results

  • monoethanolamine610814143511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Monoethanolamine

2 Products
Synonym:   2-Aminoethanol, 2-Aminoethyl alcohol, ETA, Ethanolamine, MEA 90, MEA-LCI, MEA, Monoethanolamine
Image of Monoethanolamine
CAS Number:
Linear Formula:
 NH2CH2CH2OH
Molecular Weight:
 61.08
 Beilstein/REAXYS Number:
 505944
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
Show More