• USA Home
  • Search Results

  • monomethylfumarate13010275687811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

mono-Methyl fumarate

2 Products
Synonym:   Fumaric acid monomethyl ester, Methyl hydrogen fumarate
Image of mono-Methyl fumarate
CAS Number:
Linear Formula:
 HO2CCH=CHCO2CH3
Molecular Weight:
 130.10
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More