• USA Home
  • Search Results

  • monomethylphthalate18016437618511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

mono-Methyl phthalate

2 Products
Synonym:   Monomethyl phthalate
Image of mono-Methyl phthalate
CAS Number:
Linear Formula:
 2-(HO2C)C6H4CO2CH3
Molecular Weight:
 180.16
 Beilstein/REAXYS Number:
 2048766
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More