• USA Home
  • Search Results

  • mops20926113261211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

MOPS

6 Products
Synonym:   3-(N-Morpholino)propanesulfonic acid, 4-Morpholinepropanesulfonic acid
Image of MOPS
CAS Number:
Empirical Formula (Hill Notation):
 C7H15NO4S
Molecular Weight:
 209.26
 Beilstein/REAXYS Number:
 1106776
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Absorption
  Titertest
  Solubility
  Loss
  UV-Absorption Values
  Product Line
  Special Grade
  pH Value
  UV-Absorption Condition
Show More