• USA Home
  • Search Results

  • mordantorange128723224376711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mordant Orange 1

1 Products
Synonym:   5-(4-Nitrophenylazo)salicylic acid, Alizarin Yellow R, MO1
Image of Mordant Orange 1
CAS Number:
Linear Formula:
 O2NC6H4N=NC6H3-2-(OH)CO2H
Molecular Weight:
 287.23
 Beilstein/REAXYS Number:
 962365
 Colour Index Number:
 14030
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More