• USA Home
  • Search Results

  • morphinesulfatesaltpentahydrate75883621115011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Morphine sulfate salt pentahydrate

1 Products
Synonym:   Morphine hemi[sulfate pentahydrate]
Image of Morphine sulfate salt pentahydrate
CAS Number:
Empirical Formula (Hill Notation):
 C34H40N2O10S · 5H2O
Molecular Weight:
 758.83
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Application(s)
Show More