• USA Home
  • Search Results

  • morpholine871211091811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Morpholine

5 Products
Synonym:   Tetrahydro-1,4-oxazine
Image of Morpholine
CAS Number:
Empirical Formula (Hill Notation):
 C4H9NO
Molecular Weight:
 87.12
 Beilstein/REAXYS Number:
 102549
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 39162409
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
  Potency
  Density
Show More