• USA Home
  • Search Results

  • mowiol49812345900289511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mowiol® 4-98

1 Products
Synonym:   Poly(vinyl alcohol)
Image of Mowiol<SUP>®</SUP> 4-98
CAS Number:
Linear Formula:
 [-CH2CHOH-]n
 MDL number:
 NACRES
 NA.23
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Extent of labeling
Show More

Product #
Description
81382 Mowiol® 4-98, Mw ~27,000,