• USA Home
  • Search Results

  • mttformazan335435736069711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

MTT Formazan

2 Products
Synonym:   1-(4,5-Dimethylthiazol-2-yl)-3,5-diphenylformazan, Thiazolyl blue formazan
Image of MTT Formazan
CAS Number:
Empirical Formula (Hill Notation):
 C18H17N5S
Molecular Weight:
 335.43
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Physical Form
  Application(s)
Show More