• USA Home
  • Search Results

  • mxylene1061710838311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

m-Xylene

8 Products
Synonym:   1,3-Dimethylbenzene, m-Xylene
Image of m-Xylene
CAS Number:
Linear Formula:
 C6H4(CH3)2
Molecular Weight:
 106.17
 EC Index Number
 203-576-3
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Refractive Index
  Anal. Standard Form
  Physical Form
  Transition temperature
  Titertest
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Assay
  Vapor pressure
Show More