• USA Home
  • Search Results

  • mycophenolicacid320342428093111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mycophenolic acid

3 Products
Synonym:   6-(1,3-Dihydro-7-hydroxy-5-methoxy-4-methyl-1-oxoisobenzofuran-6-yl)-4-methyl-4-hexanoic acid, 6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl)-4-methyl-4-hexenoic acid, NSC 129185
Image of Mycophenolic acid
CAS Number:
Empirical Formula (Hill Notation):
 C17H20O6
Molecular Weight:
 320.34
 Beilstein Registry Number:
 1295848
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Mode of Action
  Physical Form
  Solubility
  Melting Point
  Color
  Biological Source
  Assay
Show More