• USA Home
  • Search Results

  • myrcene1362312335311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Myrcene

5 Products
Synonym:   β-Myrcene, 7-Methyl-3-methylene-1,6-octadiene
Image of Myrcene
CAS Number:
Linear Formula:
 H2C=CHC(=CH2)CH2CH2CH=C(CH3)2
Molecular Weight:
 136.23
 Beilstein/REAXYS Number:
 1719990
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Shelf Life
  Allergen
  Organoleptic property
  Additive
  Application(s)
  Biological Source
  Assay
  Agency/Method
Show More