• USA Home
  • Search Results

  • myricitrin464381791287711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Myricitrin

4 Products
Synonym:   3,3′,4′,5,5′,7-Hexahydroxyflavone 3-O-rhamnoside, Myricetin 3-O-α-L-rhamnopyranoside, Myricetin 3-O-rhamnoside, Myricitroside
Image of Myricitrin
CAS Number:
Empirical Formula (Hill Notation):
 C21H20O12
Molecular Weight:
 464.38
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Physical Form
  Special Grade
  Manufacturer designation
  Application(s)
Show More