• USA Home
  • Search Results

  • myristicacid2283754463811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Myristic acid

9 Products
Synonym:   Tetradecanoic acid
Image of Myristic acid
CAS Number:
Linear Formula:
 CH3(CH2)12COOH
Molecular Weight:
 228.37
 EC Index Number
 208-875-2
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Anal. Standard Form
  Allergen
  Organoleptic property
  Physical Form
  Titertest
  Melting Point
  Assay
  Special Grade
Show More