• USA Home
  • Search Results

  • myristicd27acid255546065841511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Myristic-d27 acid

1 Products
Synonym:   Tetradecanoic-d27 acid
Image of Myristic-d<SUB>27</SUB> acid
CAS Number:
Linear Formula:
 CD3(CD2)12CO2H
Molecular Weight:
 255.54
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Isotopic Purity
  Application(s)
Show More