• USA Home
  • Search Results

  • myristyltrimethylammoniumbromide33639111997711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Myristyltrimethylammonium bromide

3 Products
Synonym:   Tetradecyltrimethylammonium bromide, Trimethyl(tetradecyl)ammonium bromide
Image of Myristyltrimethylammonium bromide
CAS Number:
Linear Formula:
 CH3(CH2)13N(Br)(CH3)3
Molecular Weight:
 336.39
 Beilstein/REAXYS Number:
 3633227
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Solubility
  ID clarifier
  UV-Absorption Values
Show More