• USA Home
  • Search Results

  • n2hydroxyethylethylenediaminennntriaceticacid2782615039011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic acid

2 Products
Synonym:   N-Carboxymethyl-N′-(2-hydroxyethyl)-N,N′-ethylenediglycine, HEDTA, HEEDTA
Image of <I>N</I>-<WBR>(2-<WBR>Hydroxyethyl)<WBR>ethylenediamine-<I>N</I>,<I>N</I>′,<I>N</I>′-triacetic acid
CAS Number:
Linear Formula:
 HOCH2CH2N(CH2CO2H)CH2CH2N(CH2CO2H)2
Molecular Weight:
 278.26
 Beilstein/REAXYS Number:
 1804795
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More