• USA Home
  • Search Results

  • n3aminopropyl2pyrrolidinone14220766377611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-(3-Aminopropyl)-2-pyrrolidinone

1 Products
Image of <I>N</I>-<WBR>(3-<WBR>Aminopropyl)<WBR>-<WBR>2-<WBR>pyrrolidinone
CAS Number:
Empirical Formula (Hill Notation):
 C7H14N2O
Molecular Weight:
 142.20
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More