• USA Home
  • Search Results

  • n4aminobutylnethylisoluminol276336661229111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-(4-Aminobutyl)-N-ethylisoluminol

1 Products
Synonym:   4-(N1-Ethyl-4-aminobutylamino)phthalic hydrazide, 6-[N-(4-Aminobutyl)-N-ethylamino]-2,3-dihydro-1,4-phthalazinedione, ABEI
Image of <I>N</I>-(4-Aminobutyl)-<I>N</I>-ethylisoluminol
CAS Number:
Empirical Formula (Hill Notation):
 C14H20N4O2
Molecular Weight:
 276.33
 Beilstein/REAXYS Number:
 920895
 MDL number:
 NACRES:
 NA.83
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More