• USA Home
  • Search Results

  • n62odibutyryladenosine35cyclicmonophosphatesodiumsalt491371698089511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt

2 Products
Synonym:   Bucladesine sodium salt, Dibutyryl cAMP sodium salt, Dibutyryl cyclic-AMP sodium salt
Image of N<SUP>6</SUP>,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt
CAS Number:
Linear Formula:
 C18H23N5O8PNa
Molecular Weight:
 491.37
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More