• USA Home
  • Search Results

  • nacetyldlactosamine383353218159211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Acetyl-D-lactosamine

1 Products
Synonym:   β-D-Gal-(1→4)-D-GlcNAc, 2-Acetamido-2-deoxy-4-O-β-D-galactopyranosyl-D-glucopyranose, LN, N-Acetyl-4-O-(β-D-galactopyranosyl)-D-glucosamine
Image of N-Acetyl-<SC>D</SC>-lactosamine
CAS Number:
Empirical Formula (Hill Notation):
 C14H25NO11
Molecular Weight:
 383.35
 Beilstein/REAXYS Number:
 96808
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More