• USA Home
  • Search Results

  • nacetyldpenicillamine191251553771011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Acetyl-D-penicillamine

1 Products
Synonym:   N-Acetyl-3-mercapto-D-valine, NAP, NAPA
Image of N-Acetyl-<SC>D</SC>-penicillamine
CAS Number:
Empirical Formula (Hill Notation):
 C7H13NO3S
Molecular Weight:
 191.25
 Beilstein/REAXYS Number:
 1724742
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More