• USA Home
  • Search Results

  • nacetyllcysteine1631961691111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Acetyl-L-cysteine

5 Products
Synonym:   LNAC, NAC
Image of <I>N</I>-Acetyl-<SC>L</SC>-cysteine
CAS Number:
Linear Formula:
 HSCH2CH(NHCOCH3)CO2H
Molecular Weight:
 163.19
 Beilstein/REAXYS Number:
 1724426
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Titertest
  Product Line
  Solubility
  Special Grade
Show More