• USA Home
  • Search Results

  • nacetylltyrosine2232353755311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Acetyl-L-tyrosine

3 Products
Image of <I>N</I>-Acetyl-<SC>L</SC>-tyrosine
CAS Number:
Linear Formula:
 4-(HO)C6H4CH2CH(NHCOCH3)CO2H
Molecular Weight:
 223.23
 Beilstein/REAXYS Number:
 2697172
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Assay
  Physical Form
Show More