• USA Home
  • Search Results

  • nacetylneuraminicacid3092713148611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Acetylneuraminic acid

3 Products
Synonym:   5-Acetamido-3,5-dideoxy-D-glycero-D-galactononulosonic acid, Lactaminic acid, NAN, NANA, Sialic acid
Image of N-Acetylneuraminic acid
CAS Number:
Empirical Formula (Hill Notation):
 C11H19NO9
Molecular Weight:
 309.27
 Beilstein/REAXYS Number:
 1716283
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Type
  Melting Point
  Biological Source
  Physical Form
  Application(s)
Show More