• USA Home
  • Search Results

  • nacetylneuraminylfucosyllactonneotetraosefromhumanmilk11450311121636511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Acetylneuraminyl-fucosyllacto-N-neo-tetraose from human milk

1 Products
Synonym:   α-NeuNAc-(2→6)-β-Gal-(1→4)-β-GlcNAc-(1→3)-β-Gal-(1→4)-(α-Fuc-[1→3])-Glc, SFLNnT, Sialyl-fucosyllacto-N-neo-tetraose
Image of N-<WBR>Acetylneuraminyl-<WBR>fucosyllacto-<WBR>N-<WBR>neo-<WBR>tetraose from human milk
CAS Number:
Empirical Formula (Hill Notation):
 C43H72N2O33
Molecular Weight:
 1145.03
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More