• USA Home
  • Search Results

  • nad66343anhydrousbasis5384911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

NAD

2 Products
Synonym:   β-DPN, β-NAD, β-Nicotinamide adenine dinucleotide hydrate, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide
Image of NAD
CAS Number:
Empirical Formula (Hill Notation):
 C21H27N7O14P2 · xH2O
Molecular Weight:
 663.43 (anhydrous basis)
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More