• USA Home
  • Search Results

  • naftifinehydrochloride323866547314511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Naftifine hydrochloride

2 Products
Image of Naftifine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C21H21N · HCl
Molecular Weight:
 323.86
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Mode of Action
Show More