• USA Home
  • Search Results

  • nalidixicacidsodiumsalt25422337405811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nalidixic acid sodium salt

2 Products
Synonym:   1-Ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid sodium salt
Image of Nalidixic acid sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C12H11N2NaO3
Molecular Weight:
 254.22
 Beilstein Registry Number:
 3580062
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Physical Form
  Mode of Action
Show More