• USA Home
  • Search Results

  • nallylthiourea1161810957911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Allylthiourea

2 Products
Synonym:   1-Allyl-2-thiourea, Thiosinamine
Image of <I>N</I>-Allylthiourea
CAS Number:
Linear Formula:
 CH2=CHCH2NHCSNH2
Molecular Weight:
 116.18
 Beilstein/REAXYS Number:
 1071470
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Melting Point
  Assay
Show More