• USA Home
  • Search Results

  • naloxonehydrochloridedihydrate399875148160811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Naloxone hydrochloride dihydrate

2 Products
Image of Naloxone hydrochloride dihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C19H21NO4 · HCl · 2H2O
Molecular Weight:
 399.87
 Beilstein/REAXYS Number:
 6260807
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Physical Form
Show More