• USA Home
  • Search Results

  • nanofibermultiwellplateinteractivemicroenvironmentsystemimems1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nanofiber multiwell plate, Interactive MicroEnvironMent System (IMEMS)

2 Products
Synonym:   3d plates, tissue culture plates
Image of Nanofiber multiwell plate, Interactive MicroEnvironMent System (IMEMS)
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Material
  Suitability
  Sterilization
  Matrix active group
  Manufacturer designation
  Feature
  Composite Dimension
  Color
Show More