• USA Home
  • Search Results

  • nanosilicagelontlcplates1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nano Silica Gel on TLC Plates

5 Products
Synonym:   Silica Gel Nano on TLC Plates
 NACRES:
 NC.07
 eCl@ss
 32110504
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Composite Dimension
  Quality clarifier
  Physical Form
  Special Grade
  Pore size
  Particle Dimension
  Manufacturer designation
  Application(s)
  Feature
Show More