• USA Home
  • Search Results

  • naphthalene128179120311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Naphthalene

7 Products
Image of Naphthalene
CAS Number:
Empirical Formula (Hill Notation):
 C10H8
Molecular Weight:
 128.17
 Beilstein/REAXYS Number:
 1421310
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39011601
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Shelf Life
  Melting Point
  Solubility
  Anal. Standard Form
  Titertest
  Assay
  Special Grade
Show More