• USA Home
  • Search Results

  • naphthalenebishexachlorocyclopentadieneadduct67371569692411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Naphthalene-bis(hexachlorocyclopentadiene) adduct

1 Products
Synonym:   DHA, NSC 122892
Image of Naphthalene-<WBR>bis(hexachlorocyclopentadiene) adduct
CAS Number:
Empirical Formula (Hill Notation):
 C20H8Cl12
Molecular Weight:
 673.71
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More