• USA Home
  • Search Results

  • naringin580531023647211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Naringin

5 Products
Synonym:   4′,5,7-Trihydroxyflavanone 7-rhamnoglucoside, Naringenin 7-neohesperidoside, Naringenine-7-rhamnosidoglucoside, Naringoside
Image of Naringin
CAS Number:
Empirical Formula (Hill Notation):
 C27H32O14
Molecular Weight:
 580.53
 Beilstein/REAXYS Number:
 102012
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Application(s)
  Manufacturer designation
  Physical Form
  Special Grade
Show More