• USA Home
  • Search Results

  • natosylllysinechloromethylketonehydrochloride36931427274611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nα-Tosyl-L-lysine chloromethyl ketone hydrochloride

2 Products
Synonym:   (3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, (3S)-7-Amino-1-chloro-3-tosylamino-2-heptanone hydrochloride, TLCK, Tosyl-L-lysyl-chloromethane hydrochloride
Image of <I>N</I><SUB>α</SUB>-Tosyl-<SC>L</SC>-lysine chloromethyl ketone hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C14H21ClN2O3S · HCl
Molecular Weight:
 369.31
 Beilstein/REAXYS Number:
 7106867
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More