• USA Home
  • Search Results

  • nbenzoylltyrosinepnitroanilide40540615445611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Benzoyl-L-tyrosine p-nitroanilide

1 Products
Image of N-Benzoyl-<SC>L</SC>-tyrosine p-nitroanilide
CAS Number:
Empirical Formula (Hill Notation):
 C22H19N3O5
Molecular Weight:
 405.40
 MDL number:
 PubChem Substance ID:
 eCl@ss
 32160406
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More