• USA Home
  • Search Results

  • nbenzyllprolineethylester2333195540811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Benzyl-L-proline ethyl ester

1 Products
Image of <I>N</I>-Benzyl-<SC>L</SC>-proline ethyl ester
CAS Number:
Empirical Formula (Hill Notation):
 C14H19NO2
Molecular Weight:
 233.31
 Beilstein/REAXYS Number:
 5950140
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More