• USA Home
  • Search Results

  • nbenzylquininiumchloride451006717425811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Benzylquininium chloride

1 Products
Synonym:   QUIBEC
Image of <I>N</I>-Benzylquininium chloride
CAS Number:
Empirical Formula (Hill Notation):
 C27H31ClN2O2
Molecular Weight:
 451.00
 Beilstein/REAXYS Number:
 5702637
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More