• USA Home
  • Search Results

  • nbocindole2boronicacid2610821331844611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Boc-indole-2-boronic acid

1 Products
Synonym:   1-(tert-butoxycarbonyl)indole-2-boronic acid, N-(tert-butoxycarbonyl)indole-2-boronic acid
Image of <I>N</I>-Boc-indole-2-boronic acid
CAS Number:
Empirical Formula (Hill Notation):
 C13H16BNO4
Molecular Weight:
 261.08
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More