• USA Home
  • Search Results

  • nboclprolinol201266961040811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Boc-L-prolinol

1 Products
Synonym:   (S)-1-Boc-2-pyrrolidinemethanol
Image of <I>N</I>-Boc-<SC>L</SC>-prolinol
CAS Number:
Empirical Formula (Hill Notation):
 C10H19NO3
Molecular Weight:
 201.26
 Beilstein/REAXYS Number:
 3542667
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
446327 N-Boc-L-prolinol, 98%,