• USA Home
  • Search Results

  • nbromosuccinimide1779812808511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Bromosuccinimide

2 Products
Synonym:   NBS
Image of <I>N</I>-Bromosuccinimide
CAS Number:
Empirical Formula (Hill Notation):
 C4H4BrNO2
Molecular Weight:
 177.98
 Beilstein/REAXYS Number:
 113916
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Melting Point
Show More