• USA Home
  • Search Results

  • nbutyllithiumsolution640610972811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

n-Butyllithium solution

6 Products
Synonym:   n-BuLi, Butyl lithium, Butyllithium solution, Lithium-1-butanide
Image of <I>n</I>-Butyllithium solution
CAS Number:
Linear Formula:
 CH3(CH2)3Li
Molecular Weight:
 64.06
 Beilstein/REAXYS Number:
 1209227
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Shelf Life
  Color
  Physical Form
  Density
Show More