• USA Home
  • Search Results

  • ndecanoylnmethylglucamine349468526120711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Decanoyl-N-methylglucamine

1 Products
Synonym:   N-(D-Glucityl)-N-methyldecanamide, N-Decanoyl-N-methyl-D-glucamine, MEGA-10
Image of <I>N</I>-Decanoyl-<I>N</I>-methylglucamine
CAS Number:
Empirical Formula (Hill Notation):
 C17H35NO6
Molecular Weight:
 349.46
 Beilstein/REAXYS Number:
 6974359
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More