• USA Home
  • Search Results

  • ndodecylbdmaltoside510626922793611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

n-Dodecyl β-D-maltoside

2 Products
Synonym:   DDM, Lauryl-β-D-maltoside
Image of <I>n</I>-Dodecyl β-<SC>D</SC>-maltoside
CAS Number:
Empirical Formula (Hill Notation):
 C24H46O11
Molecular Weight:
 510.62
 Beilstein/REAXYS Number:
 55318
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
 eCl@ss:
 32190102
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More