• USA Home
  • Search Results

  • ndodecylnndimethyl3ammonio1propanesulfonate335551493308511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate

2 Products
Synonym:   3-(N,N-Dimethyldodecylammonio)propanesulfonate, 3-(N,N-Dimethyllaurylammonio)propanesulfonate, 3-(Dodecyldimethylammonio)propanesulfonate, 3-(Lauryldimethylammonio)propanesulfonate, Dodecyldimethyl(3-sulfopropyl)ammonium hydroxide inner salt, Lauryl sulfobetaine, SB3-12
Image of <I>N</I>-Dodecyl-<I>N</I>,<I>N</I>-<WBR>dimethyl-<WBR>3-<WBR>ammonio-<WBR>1-<WBR>propanesulfonate
CAS Number:
Linear Formula:
 CH3(CH2)11N+(CH3)2CH2CH2CH2SO3-
Molecular Weight:
 335.55
 Beilstein/REAXYS Number:
 4145308
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
 eCl@ss:
 32190102
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  ID clarifier
Show More