• USA Home
  • Search Results

  • neochlorogenicacid3543190633211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Neochlorogenic acid

3 Products
Synonym:   5-O-(trans-3,4-Dihydroxycinnamoyl)-D-quinic acid, trans-5-O-Caffeoylquinic acid
Image of Neochlorogenic acid
CAS Number:
Empirical Formula (Hill Notation):
 C16H18O9
Molecular Weight:
 354.31
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More